Läs mer
H-Pro-Glu-Oh, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | H-Pro-Glu-Oh |
| CAS | 67644-00-2 |
| Molekylformel | C10 H16 N2 O5 |
| Kvantitet | 100 mg |
| Synonym | L-Glutamic acid','L-prolyl- (9CI','ACI), L-Glutamic acid','N-L-prolyl- (ZCI), L-Prolyl-L-glutamic acid (ACI), 107: PN: US20130123467 SEQID: 130 claimed protein, 11: PN: WO2014063098 TABLE: 5 claimed protein, 459: PN: WO2005081628 SEQID: 1568 claimed protein, 6: PN: CN109305994 PAGE: 5 claimed sequence |
| InChI formel | InChI=1S/C10H16N2O5/c13-8(14)4-3-7(10(16)17)12-9(15)6-2-1-5-11-6/h6-7,11H,1-5H2,(H,12,15)(H,13,14)(H,16,17)/t6-,7-/m0/s1 |
| LEDER | OC(=O)CC[C@H](NC(=O)[C@@H]1CCCN1)C(=O)O |
| IUPAC-namn | (2S)-2-[[(2S)-pyrrolidine-2-carbonyl]amino]pentanedioic acid |
| Molekylvikt (g/mol) | 244.244 |
| Formel vikt | 244.1059 |