Läs mer
H-Pro-NHEt Hydrochloride, TRC
Produktkod. 30424868
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424868 100 mg
30421074 10 mg
30416911 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424868 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP839230100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58107-62-3
Molekylformel C7H14N2O . HCl
Molekylvikt (g/mol) 178.66
IUPAC-namn (2S)-N-ethylpyrrolidine-2-carboxamide;hydrochloride
LEDER Cl.CCNC(=O)[C@@H]1CCCN1
Specifikationer
Kemiskt namn eller material H-Pro-NHEt Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C7H14N2O.ClH/c1-2-8-7(10)6-4-3-5-9-6;/h6,9H,2-5H2,1H3,(H,8,10);1H/t6-;/m0./s1
Formel vikt 178.0873