Läs mer
H-Trp-Phe-OH, TRC
Produktkod. 30406922
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30406922 500 mg
1 options
Produktkod. 30406922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH218605500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material H-Trp-Phe-OH
CAS 6686-02-8
Molekylformel C20 H21 N3 O3
Kvantitet 500 mg
Synonym L-Tryptophyl-L-phenylalanine, 14: PN: JP2014079213 TABLE: 2 claimed sequence, 2: PN: WO2013070776 PAGE: 109 claimed sequence, Tryptophanylphenylalanine, Tryptophylphenylalanine
InChI formel InChI=1S/C20H21N3O3/c21-16(11-14-12-22-17-9-5-4-8-15(14)17)19(24)23-18(20(25)26)10-13-6-2-1-3-7-13/h1-9,12,16,18,22H,10-11,21H2,(H,23,24)(H,25,26)/t16-,18-/m0/s1
LEDER N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](Cc3ccccc3)C(=O)O
IUPAC-namn (2S)-2-[[(2S)-2-amino-3-(1H-indol-3-yl)propanoyl]amino]-3-phenylpropanoic acid
Molekylvikt (g/mol) 351.399
Formel vikt 351.158