Läs mer
H-Tyr-Val-OH, TRC
Produktkod. 30456536
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456536 100 mg
30454066 10 mg
30449461 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30456536 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT966410100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17355-09-8
Molekylformel C14H20N2O4
Molekylvikt (g/mol) 280.32
IUPAC-namn (2S)-2-[[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]amino]-3-methylbutanoic acid
LEDER CC(C)[C@H](NC(=O)[C@@H](N)Cc1ccc(O)cc1)C(=O)O
Specifikationer
Kemiskt namn eller material H-Tyr-Val-OH
InChI formel InChI=1S/C14H20N2O4/c1-8(2)12(14(19)20)16-13(18)11(15)7-9-3-5-10(17)6-4-9/h3-6,8,11-12,17H,7,15H2,1-2H3,(H,16,18)(H,19,20)/t11-,12-/m0/s1
Kvantitet 100 mg
Formel vikt 280.1423