Learn More
Erlab™ H14 HEPA Filter
Erlab™ H14 HEPA Filter can be use with a range of filtrating fume hoods and storage cabinets.
Brand:  Erlab™ H01074031
Additional Details : Weight : 1.00000kg
Product Code. 16485229
  • 6683.30 SEK / Each
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
  • Filter HP - HEPA H14 : This filtration technology traps particles larger than 0.1 μm with 99.995
  • For use with powders
Specifications
Specifications
GF4/HP
Filter
Powders