missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ H1FOO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558200
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: YRKTKAESKS SKPTASKVKN GAASPTKKKV VAKAKAPKAG QGPNTKAAAP AKGSGSKVVP AHLSRKTEAP KGPRKAGLPI KASSSKVSSQ Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 36%.
Eukaryotic Histones are basic and water soluble nuclear proteins that form hetero-octameric nucleosome particles by wrapping 146 base pairs of DNA in a left-handed super-helical turn sequentially to form chromosomal fibers. Two molecules of each of the four core Histones (Histone H2A, H2B, H3, and H4) form the octamer, which consists of two H2A-H2B dimers and two H3-H4 dimers that are nearly symmetrical by tertiary structure. Over 80% of nucleosomes contain the linker Histone H1, derived from an intronless gene, that interacts with linker DNA between nucleosomes and mediates compaction into higher order chromatin. H1FOO (H1 histone family, member O, oocyte-specific), also known as OSH1, is a 346 amino acid oocyte-specific Histone that localizes to both the nucleus and the cytoplasm. Expressed as multiple alternatively spliced isoforms, H1FOO is thought to play an important role in gene control during oogenesis and early embryogenesis and is crucial for meiotic maturation of germinal vesicle-stage oocytes.
Specifikationer
| H1FOO | |
| Polyclonal | |
| Unconjugated | |
| H1FOO | |
| C86609; H1 histone family member O, oocyte specific; H1 histone family, member O, oocyte-specific; H1.8; H1fo; H1foo; H1OO; Histone H1oo; oocyte-specific histone H1; Oocyte-specific linker histone H1; OSH1; RGD1566236 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 132243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZA3 | |
| H1FOO | |
| Recombinant protein corresponding to Human H1FOO. Recombinant protein control fragment (Product #RP-96499). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering