missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ H1N1 (A/South Carolina/1/18, AAD17229, 19 a.a. - 344 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Brand: Abnova™ P4895.50ug
Additional Details : Weight : 0.02000kg
Specifications
AAD17229 | |
SDS-PAGE | |
H1N1 (A/South Carolina/1/18) Recombinant Protein | |
50 ug | |
RUO | |
Human | |
His | |
Liquid |
1 ug/uL | |
Liquid | |
Mammalian cell (293) expression system | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
H1N1 | |
Recombinant | |
Mammalian cell (293) expression system | |
>95% by SDS-PAGE |