missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2AFY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17030-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Variant histone H2A which replaces conventional H2A in a subset of nucleosomes where it represses transcription. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. May be involved in stable X chromosome inactivation.Specifications
H2AFY2 | |
Polyclonal | |
Unconjugated | |
H2AFY2 | |
Core histone macro H2A.2, H2A histone family, member Y2, H2AFY2, Histone macroH2A2, macroH2A2, mH2A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361844, 404634, 55506 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CCK0, Q9P0M6 | |
H2afy2 | |
H2AFY2 Fusion Protein Ag10487 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |