Läs mer
MedChemExpress H2N-PEG2-CH2COOH
Produktkod. 16419326
Encompass_Preferred
Change view
100 mg
Produktkod.
16419326 100 mg
1 options
Produktkod. 16419326 Leverantör MedChemExpress Leverantörsnummer HYW006524
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
MedChemExpress H2N-PEG2-CH2COOH is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.
Specifikationer
Molekylvikt (g/mol) 163.17 g/mol
CAS 134978-97-5
Innehåll och lagring Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Frakt skick Room temperature
Kvantitet 100 mg
Mål NO Synthase ∣ MMP ∣ Endogenous Metabolite