missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ H4ac pan-acetyl (K5,K8,K12) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540083
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
H4ac pan-acetyl (K5,K8,K12) Polyclonal Antibody for Western Blot, ICC/IF, ELISA, ChIP, ChIP-seq, ARRAY
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifikationer
| H4ac pan-acetyl (K5,K8,K12) | |
| Polyclonal | |
| Unconjugated | |
| H4C1 | |
| acetyl Histone H4; Acetyl-H4 K5 K8 K12; Acetyl-H4 Lys5 Lys8 Lys12; Acetyl-Histone H4 K5 K8 K12; Acetyl-Histone H4 Lys5 Lys8 Lys12; FO108; H4; H4 ac pan-acetyl; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/j; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4ac; H4C1; H4C11; H4c12; H4C13; H4c14; H4C15; H4C2; H4C3; H4c4; H4C5; H4C6; H4c8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K12; H4K12a; H4K12ac; H4K5; H4K5,8,12ac; H4K5ac; H4K8; H4K8ac; H4Lys5,8,12ac; H4M; HIST1H4; Hist1h4a; Hist1h4b; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; Hist1h4h; Hist1h4i; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; HIST2H4; HIST2H4A; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; histone H4; Histone H4 acetyl; Histone H4A; Histone H4K12; Histone H4K5; Histone H4K8; histone IV, family 2; X04652 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121504, 319158, 326619, 326620, 8294, 8359, 8363, 8366, 8367, 8370, 97122 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Peptide Array, ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Western Blot, Immunocytochemistry | |
| 1.05 mg/mL | |
| PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 | |
| P62805, P62806 | |
| H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9 | |
| KLH-conjugated synthetic peptide corresponding to human the region of histone H4 containing the acetylated lysines 5, 8 and 12 (H4K5,8,12ac). | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering