Läs mer
Thermo Scientific™ HAAKE™ Viscotester™ 1 Plus and 2 Plus Rotational Viscometers
Handla allt Thermo Scientific ProdukterBeskrivning
The HAAKE Visctoester 1, 2 plus Viscometer has been discontinued and replaced with the Thermo Scientific™ HAAKE™ Viscotester 3 Viscometer.
We continue to offer a comprehensive portfolio of accessories, spare parts and services for the The HAAKE Visctoester 1, 2 plus Viscometer to support your daily work
The HAAKE Visctoester 1, 2 plus Viscometer has been discontinued and replaced with the Thermo Scientific™ HAAKE™ Viscotester 3 Viscometer.
We continue to offer a comprehensive portfolio of accessories, spare parts and services for the The HAAKE Visctoester 1, 2 plus Viscometer to support your daily work.
Specifikationer
Specifikationer
| Produkttyp | Rotational Viscometer |
| Viskositetsområde | 1,5 till 330 mPas |
| Beskrivning | This product has been discontinued and replaced with the Thermo Scientific™ HAAKE™ Viscotester 3 Viscometer. |
| Noggrannhet | ±5% (Standard), ±1% (Optional) |
| Reproducerbarhet | ±1% (FSD) |