Läs mer
Tocris Bioscience™ HADA
Beskrivning
HADA is a fluorescent D-amino acid. Suitable for labeling peptidoglycans in live bacteria. Results in strong peripheral and septal labeling of diverse bacterial cell populations without affecting growth rates. Displays mitochondrial outer membrane permeability. Excitation/emission λ ∽405/450 nm.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-[[(7-Hydroxy-2-oxo-2H-1-benzopyran-3-yl)carbonyl]amino]-D-alanine hydrocholoride |
| CAS | 2253733-10-5 |
| Kvantitet | 5 mg |
| Molekylformel | C13H12N2O6.HCl |
| Formel vikt | 328.71 |
| Renhet | 98% |