Learn More
Proteintech HADHB Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29091-1-AP-20UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17231426
  • 1355.00 SEK / 20µL
Estimated Shipment: 08-06-2023
to see stock.
Description
Description
The HADHB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HADHB. This antibody recognizes human, mouse, rat antigen. The HADHB antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
HADHB
Polyclonal
Unconjugated
20 μL
3032
Liquid
Western Blot, ELISA
350 μg/mL
Rabbit
Primary
Human, Mouse, Rat