Learn More
CleanSpace Technology™ HALO Particulate Filter P3 Standard
Suitable for protection against airborne particulates (dust, mists and fumes) and biohazards.
Brand:  CleanSpace Technology™ CS3002
143.65 SEK valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Additional Details : Weight : 0.11970kg
Product Code. 15980032
  • 1360.00 SEK / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Suitable for protection against airborne particulate (dust, mists and fumes).
  • Materials: Fiberglass particulate media and plastic casing, silicone seal
  • Easy and quickly fitted and removed from the power unit
  • Pack of 3 (CS3002)
Specifications
Specifications
Filter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
IMPORTANT: The filters are compatible with CleanSpace HALO Power Units. When selecting a CleanSpace Filter please consult a Health and Safety specialist for advice on the appropriate respiratory equipment and filter use.