Learn More
CleanSpace Technology™ HALO Particulate Filter P3 Standard
Suitable for protection against airborne particulates (dust, mists and fumes) and biohazards.
Brand: CleanSpace Technology™ CS3002
143.65 SEK valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Additional Details : Weight : 0.11970kg
Description
- Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
- Suitable for protection against airborne particulate (dust, mists and fumes).
- Materials: Fiberglass particulate media and plastic casing, silicone seal
- Easy and quickly fitted and removed from the power unit
- Pack of 3 (CS3002)
Specifications
Filter |
IMPORTANT: The filters are compatible with CleanSpace HALO Power Units. When selecting a CleanSpace Filter please consult a Health and Safety specialist for advice on the appropriate respiratory equipment and filter use.