Läs mer
Haloperidol pyridinium Chloride Impurity, TRC
Produktkod. 30136046
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136046 50 mg
30007729 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136046 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH10377050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 125785-69-5
Molekylformel C21H18Cl2FNO
Molekylvikt (g/mol) 390.28
Synonym HPP+ ion Chloride, 4-(4-Chlorophenyl)-1-(4-(4-fluorophenyl)-4-oxobutyl)pyridin-1-ium Chloride
LEDER O=C(CCC[N+]1=CC=C(C2=CC=C(Cl)C=C2)C=C1)C3=CC=C(F)C=C3.[Cl-]
Specifikationer
Kemiskt namn eller material Haloperidol Pyridinium Chloride Impurity
Kvantitet 50 mg
InChI formel InChI=1S/C21H18ClFNO/c22-19-7-3-16(4-8-19)17-11-14-24(15-12-17)13-1-2-21(25)18-5-9-20(23)10-6-18/h3-12,14-15H,1-2,13H2/q+1
Formel vikt 354.1061