Läs mer
Hamamelitannin, TRC
Produktkod. 30004372
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30004372 1 mg -
30006083 0.5 mg 0.5mg
30004552 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004372 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH1754601MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 469-32-9
Molekylformel C20 H20 O14
Molekylvikt (g/mol) 484.36
Synonym D-Ribose, 2-C-[[(3,4,5-trihydroxybenzoyl)oxy]methyl]-, 5-(3,4,5-trihydroxybenzoate), Hamamelitannin (6CI,7CI), Ribose, 2-C-(hydroxymethyl)-, 21,5-digallate, D- (8CI), Gallic acid, 21,5-diester with 2-C-(hydroxymethyl)-D-ribose (8CI), 2',5-Di-O-galloyl-D-hamamelose
IUPAC-namn [(2S,3S,4S)-3,4,5-trihydroxy-4-[(3,4,5-trihydroxybenzoyl)oxymethyl]oxolan-2-yl]methyl 3,4,5-trihydroxybenzoate
LEDER OC1O[C@@H](COC(=O)c2cc(O)c(O)c(O)c2)[C@H](O)[C@@]1(O)COC(=O)c3cc(O)c(O)c(O)c3
Specifikationer
Kemiskt namn eller material Hamamelitannin
Kvantitet 1 mg
InChI formel InChI=1S/C20H20O14/c21-9-1-7(2-10(22)14(9)25)17(28)32-5-13-16(27)20(31,19(30)34-13)6-33-18(29)8-3-11(23)15(26)12(24)4-8/h1-4,13,16,19,21-27,30-31H,5-6H2/t13-,16-,19?,20-/m0/s1
Formel vikt 484.0853