Learn More
Hamilton™ 1800 Series Gastight™ Replacement Syringe Barrels
Barrels range in volume from 10μ to 250μL with removable needle (RN) and cemented needle (N) terminations.
835.00 SEK
Specifications
Material | PTFE |
---|---|
For Use With (Application) | Gases and liquids |
Product Type | Replacement Syringe Barrel |
Product Code | Brand | For Use With (Equipment) | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Volume (Metric) | Price | Quantity & Availability | ||||
15573963
|
Hamilton™
32190 |
1810 RN Syringe | 100 μL |
835.00 SEK
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Specifications
PTFE | |
Gases and liquids | |
Replacement Syringe Barrel |