Learn More
Hamilton™ Replacement Needles for Agilent 7673, 7683, 7693, and 6850 ALS GC Syringes
Can be used with removable (RN)-style syringes that are 100 μL and smaller. These needles are manufactured for use with Agilent Instrumentation 7673-7683, 6850, 7670, 7671, and 7672 ALS models. Custom lengths and point styles are available.
Brand: Hamilton™ 7786-01
Additional Details : Weight : 0.01000kg
Description
Needles are available in 23s gauge, 26s gauge, and 23s-26s gauge.
Six needles per pack.
Specifications
Autoclavable | |
1.69 in. | |
Removable AS |
23s G | |
Replacement Needle | |
Agilent 7673-7683, 6850, 7670, 7671 and 7672 ALS |