Learn More
Hamilton™ Replacement Needles for Agilent 7673, 7683, 7693, and 6850 ALS GC Syringes
Can be used with removable (RN)-style syringes that are 100 μL and smaller. These needles are manufactured for use with Agilent Instrumentation 7673-7683, 6850, 7670, 7671, and 7672 ALS models. Custom lengths and point styles are available.
Brand:  Hamilton™ 7785-01
Additional Details : Weight : 0.02500kg
 View more versions of this product
Product Code. 11552265
  • 1080.00 SEK / Pack of 6
Estimated Shipment: 29-04-2024
to see stock.
Description
Description
Needles are available in 23s gauge, 26s gauge, and 23s-26s gauge.
Six needles per pack.
Specifications
Specifications
Autoclavable
1.69 in.
Removable AS
23s to 26s G
Replacement Needle
Agilent 7673, 7683, 7693A and 6850 Autosamplers