Learn More
Hamilton™ Replacement Needles for Agilent 7673, 7683, 7693, and 6850 ALS GC Syringes
Can be used with removable (RN)-style syringes that are 100 μL and smaller. These needles are manufactured for use with Agilent Instrumentation 7673-7683, 6850, 7670, 7671, and 7672 ALS models. Custom lengths and point styles are available.
1070.00 SEK - 1080.00 SEK
Specifications
Autoclavable | Autoclavable |
---|---|
Length (English) Needle | 1.69 in. |
Product Type | Replacement Needle |
Needle Point Style | Removable AS |
Product Code | Brand | For Use With (Equipment) | Needle Gauge | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Needle Gauge | Price | Quantity & Availability | ||||
11547884
|
Hamilton™
7786-01 |
Agilent 7673-7683, 6850, 7670, 7671 and 7672 ALS | 23s G |
1070.00 SEK
Pack of 6 |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11552265
|
Hamilton™
7785-01 |
Agilent 7673, 7683, 7693A and 6850 Autosamplers | 23s to 26s G |
1080.00 SEK
Pack of 6 |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11552235
|
Hamilton™
7786-02 |
Agilent 7673-7683, 6850, 7670, 7671 and 7672 ALS | 26s G |
1070.00 SEK
Pack of 6 |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Needles are available in 23s gauge, 26s gauge, and 23s-26s gauge.
Six needles per pack.
Specifications
Autoclavable | |
1.69 in. | |
Replacement Needle | |
Removable AS |