missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Checker™ pH Tester and Replacement Part
For use with virtually any electrode that shares same connector
548.68 SEK - 665.00 SEK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12333408 View accessory products |
Hanna Instruments™
CHECKER1 |
pH Tester |
665.00 SEK Each |
Remote Stock (10 days availability): 44 |
10131361 |
Hanna Instruments™
HI98103 |
pH Tester |
548.68 SEK Each |
|
Description
- pH range from 0.00 to 14.00
- Provides high accuracy with 0.01 pH resolution at 0° to 50°C
- Accuracy is ±0.2 pH
- 1.5V
- RH max 95%
- With LCD and manual two point calibration
- 3000 hours of battery life
Specifications
pH |