missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAPLN2 Rabbit anti-Human, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27818-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
BRAL1 is a member a superfamily consisting of several highly homologous hyaluro and proteoglycan binding link proteins. BRAL1 is predomitly expressed in brain tissue and spinal cord. Like other members in the link-module superfamily, BRAL1 contains an immunoglobulin-like fold and two proteoglycan tandem repeats (PTRs). Its mRNA expression pattern is similar to other lectican proteoglycans, suggesting that BRAL1 may act to stabilize the binding between the extracellular matrix molecule hyaluro and brevican. Immunostaining of mouse brain showed BRAL1 expression at P20 in the white matter of the developing cerebellum and in myelinated fiber tracts in the adult brain, suggesting that expression starts when axonal myelination occurs.Specifications
HAPLN2 | |
Polyclonal | |
Unconjugated | |
HAPLN2 | |
Brain link protein 1, BRAL1, HAPLN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100628124, 60484, 64057 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ESM2, Q9GZV7 | |
HAPLN2 | |
HAPLN2 Fusion Protein Ag27169 | |
150 μL | |
Primary | |
Human, Porcine, Rat | |
Antibody | |
IgG |