Learn More
RETSCH Hard Anodized Aluminum Dividing Heads
Brand:  RETSCH 42.793.0002
434283.00 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 5.90000kg
 View more versions of this product
Product Code. 10004350
  • 457140.00 SEK / Each
Estimated Shipment: 02-07-2024
to see stock.
Specifications
Dividing Head
For Feed Sizes up to 10mm
10 Quick-release Sample Outlets
Hard Anodized Aluminum