Learn More
RETSCH Hard Anodized Aluminum Dividing Heads
42885.00 SEK - 457140.00 SEK
Product Code | Brand | Includes | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | For Use With (Equipment) | Price | Quantity & Availability | ||||
10004350
|
RETSCH
42.793.0002 |
10 Quick-release Sample Outlets | For Feed Sizes up to 10mm |
457140.00 SEK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
10762711
|
RETSCH
42.793.0009 |
8 Quick-release Sample Outlets | For Feed Sizes up to 5mm |
46605.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
10004360
|
RETSCH
42.793.0003 |
6 Quick-release Sample Outlets | For Feed Sizes up to 10mm |
42885.00 SEK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
10044370
|
RETSCH
42.793.0001 |
8 Quick-release Sample Outlets | For Feed Sizes up to 10mm |
44740.00 SEK
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
Specifications
Dividing Head | |
Hard Anodized Aluminum |