Learn More
Harmaline, 98+%
Product Code. 15414249
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414249 5 g 5g
15404249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15414249 Supplier Thermo Scientific Alfa Aesar Supplier No. J61699.06
 Restricted Item
Beräknad leverans: 11-12-2026
Add to basket
CAS: 304-21-2 | C13H14N2O | 214.268 g/mol
CNS stimulant; may act through NMDA receptors. Reversible inhibitor of MAO-A
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • CNS stimulant; may act through NMDA receptors
  • Reversible inhibitor of MAO-A
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 304-21-2
Molecular Formula C13H14N2O
Molecular Weight (g/mol) 214.268
MDL Number MFCD00004955
InChI Key QJOZJXNKVMFAET-UHFFFAOYSA-N
Synonym harmaline, dihydroharmine, harmidine, armalin, harmalol methyl ether, o-methylharmalol, 3,4-dihydroharmine, harmine, dihydro, harmalin, 1-methyl-7-methoxy-3,4-dihydro-beta-carboline
PubChem CID 5280951
IUPAC Name 7-methoxy-1-methyl-3,4-dihydro-2H-pyrido[3,4-b]indole
SMILES CC1=C2C(=C3C=CC(=CC3=N2)OC)CCN1
Specifications
Melting Point 239°C to 241°C
Color Brown
Quantity 5 g
Beilstein 207310
Merck Index 14,4613
Formula Weight 214.27
Percent Purity ≥98%
Physical Form Crystals
Chemical Name or Material Harmaline