Läs mer
Harman, TRC
Produktkod. 30136876
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136876 100 mg
17987914 5 mg
17999974 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136876 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH105000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 486-84-0
Molekylformel C12 H10 N2
Molekylvikt (g/mol) 182.22
Synonym 2,9-Pyridindole, 1-methyl- (3CI), 1-9H-Pyrido[3,4-b]indole, 1-methyl-, Methyl-9H-pyrido[3,4-b]indole, 1-Methyl-β-carboline, Aribin, Aribine, Harman, Harmane, Indoter, Locuturin, Locuturine, NSC 54439, Zygofabagine
IUPAC-namn 1-methyl-9H-pyrido[3,4-b]indole
LEDER Cc1nccc2c3ccccc3[nH]c12
Specifikationer
Kemiskt namn eller material Harmane
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H10N2/c1-8-12-10(6-7-13-8)9-4-2-3-5-11(9)14-12/h2-7,14H,1H3
Formel vikt 182.0844
Procent renhet >95%