Läs mer
Harmol Hydrochloride Dihydrate, TRC
Produktkod. 30397627
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30397627 1 g
1 options
Produktkod. 30397627 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH1052601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Harmol hydrochloride dihydrate
CAS 149022-16-2
Molekylformel C12 H10 N2 O . H Cl . 2 H2 O
Kvantitet 1 g
Synonym 1-Methyl-9H-pyrido(3,4-b)indol-7-ol monohydrochloride dihydrate
InChI formel InChI=1S/C12H10N2O.ClH.2H2O/c1-7-12-10(4-5-13-7)9-3-2-8(15)6-11(9)14-12;;;/h2-6,14-15H,1H3;1H;2*1H2
LEDER O.O.Cl.Cc1nccc2c3ccc(O)cc3[nH]c12
IUPAC-namn 1-methyl-9H-pyrido[3,4-b]indol-7-ol;dihydrate;hydrochloride
Molekylvikt (g/mol) 270.71
Formel vikt 270.0771