Läs mer
Harpagoside, TRC
Produktkod. 30136345
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136345 100 mg
30003673 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH105275100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19210-12-9
Molekylformel C24 H30 O11
Molekylvikt (g/mol) 494.49
Synonym (1S,4aS,5R,7S,7aS)-1,4a,5,6,7,7a-Hexahydro-4a,5-dihydroxy-7-methyl-7-[[(2E)-1-oxo-3-phenyl-2-propen-1-yl]oxy]cyclopenta[c]pyran-1-yl-β-D-glucopyranoside, Harpagoside, [1S-[1α,4aα,5α,7α(E),7aα]]-1,4a,5,6,7,7a-Hexahydro-4a,5-dihydroxy-7-methyl-7-[(1-oxo-3-phenyl-2-propenyl)oxy]cyclopenta[c]pyran-1-yl-β-D-glucopyranoside, E-Harpagoside
IUPAC-namn [(1S,4aS,5R,7S,7aS)-4a,5-dihydroxy-7-methyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,5,6,7a-tetrahydrocyclopenta[c]pyran-7-yl] (E)-3-phenylprop-2-enoate
LEDER C[C@@]1(C[C@@H](O)[C@]2(O)C=CO[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H]12)OC(=O)\C=C\c4ccccc4
Specifikationer
Kemiskt namn eller material Harpagoside
Kvantitet 100 mg
InChI formel InChI=1S/C24H30O11/c1-23(35-16(27)8-7-13-5-3-2-4-6-13)11-15(26)24(31)9-10-32-22(20(23)24)34-21-19(30)18(29)17(28)14(12-25)33-21/h2-10,14-15,17-22,25-26,28-31H,11-12H2,1H3/b8-7+/t14-,15-,17-,18+,19-,20-,21+,22+,23+,24-/m1/s1
Formel vikt 494.1788