missing translation for 'onlineSavingsMsg'
Learn More

HARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11301-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828283

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody can bind both HARSs for the close sequences.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes likely share a bidirectional promoter. Mutations in this gene are associated with the pathogenesis of Perrault syndrome, which involves ovarian dysgenesis and sensorineural hearing loss. Alternative splicing results in multiple transcript variants of this gene.
Specifications

Specifications

HARS2
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49590, Q99KK9
HARS2, HARSL, HARSR, HisRS, Histidine tRNA ligase, Histidine tRNA ligase like, HO3
Rabbit
IgG
20 μL
-20°C
Primary
23438, 307491, 70791
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
HARS2
Liquid
Hars2
HARS2 Fusion Protein Ag1827
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers