Learn More
Hastelloy C™ gauze, 10 mesh woven from 0.64mm (0.025 in.) dia. wire, Thermo Scientific™
432.00 SEK - 3248.00 SEK
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15457255
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046563.FL
75 x 75 mm
432.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
15467255
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046563.GT
150 x 150 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15477255
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046563.HB
300 x 300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products