missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HBEGF (Human) Recombinant Protein
Human HBEGF full-length ORF ( AAH33097.1, 20 a.a. - 208 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001839-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: LVTGESLERLRRGLAAGTSNPDPPTVSTDQLLPLGGGRDRKVRDLQEADLDLLRVTLSSKPQALATPNKEEHGKRKKKGKGLGKKRDPCLRKYKDFCIHGECKYVKELRAPSCICHPGYHGERCHGLSLPVENRLYTYDHTTILAVVAVVLSSVCLLVIVGLLMFRYHRRGGYDVENEEKVKLGMTNSH
Specifications
AAH33097.1 | |
heparin-binding EGF-like growth factor | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBEGF | |
GST |
1839 | |
Wheat germ expression system | |
10 μg | |
DTR, DTS, DTSF, HEGFL | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |