missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCFC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13567-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described.Specifications
HCFC2 | |
Polyclonal | |
Unconjugated | |
HCFC2 | |
C2 factor, HCF 2, HCF2, HCFC2, Host cell factor 2, host cell factor C2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29915, 314704, 67933 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5RKG2, Q9D968, Q9Y5Z7 | |
Hcfc2 | |
HCFC2 Fusion Protein Ag4461 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |