missing translation for 'onlineSavingsMsg'
Learn More

HCN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13745-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822124

  • 1656.73 SEK / 20µL
Estimated Shipment: 03-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HCN3 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. HCN3 has been detected in the brain, but with less expression than other HCN channels.
Specifications

Specifications

HCN3
Polyclonal
Unconjugated
HCN3
HCN3, KIAA1535
Rabbit
Antigen Affinity Chromatography
RUO
114245, 15168, 57657
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88705, Q9JKA8, Q9P1Z3
HCN3
HCN3 Fusion Protein Ag4684
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers