missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13745-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
HCN3 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. HCN3 has been detected in the brain, but with less expression than other HCN channels.Specifications
HCN3 | |
Polyclonal | |
Unconjugated | |
HCN3 | |
HCN3, KIAA1535 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114245, 15168, 57657 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88705, Q9JKA8, Q9P1Z3 | |
HCN3 | |
HCN3 Fusion Protein Ag4684 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |