missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
HCoV-NL63 Human Coronavirus Spike RBD Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11023-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Spike RBD Monoclonal antibody specifically detects Spike RBD in HCoV-NL63 samples. It is validated for Immunocytochemistry
Specifikationer
| Spike RBD | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Immunocytochemistry | |
| 2841A | |
| Immunocytochemistry 3-25 ug/mL | |
| YP_003767.1 | |
| Human embryonic kidney cell HEK293-derived HCoV-NL63 Spike RBD protein, Ala475-Asp634, Accession # YP_003767.1 | |
| 100 μg | |
| Primary | |
| HCoV-NL63 | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering