missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10197-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human HDAC1. The calculated molecular weight of HDAC1 is 55 kDa, but modified HDAC1 is about 60-65 kDa.
Histone deacetylase (HDAC) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones - H2A, H2B, H3, and H4. Histone deacetylation gives a tag for epigenetic repression and plays and important role in transcriptional regulation, cell cycle progression, and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.Specifications
HDAC1 | |
Polyclonal | |
Unconjugated | |
HDAC1 | |
GON 10, HD1, HDAC1, histone deacetylase 1, RPD3, RPD3L1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297893, 3065, 433759 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Flow Cytometry | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O09106, Q13547, Q4QQW4 | |
Hdac1 | |
HDAC1 Fusion Protein Ag0256 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |