missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC10 Mouse anti-Human, Unconjugated, Clone: 1A1B11, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HDAC10 |
---|---|
Clone | 1A1B11 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800396
|
Proteintech
67646-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16890386
|
Proteintech
67646-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The HDAC10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC10. This antibody recognizes human, mouse, rat antigen. The HDAC10 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
HDAC10 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
83933 | |
Primary |
1A1B11 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |