Learn More
HDAC9 Mouse anti-Human, Unconjugated, Clone: 1G6C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67364-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The HDAC9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC9. This antibody recognizes human antigen. The HDAC9 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
HDAC9 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1G6C5 | |
Unconjugated | |
150 μL | |
9734 | |
IgG2a |