missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDLBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15406-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
HDLBP | |
Polyclonal | |
Unconjugated | |
HDLBP | |
HBP, HDL binding protein, HDLBP, PRO2900, VGL, Vigilin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110611, 3069, 64474 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q00341, Q8VDJ3, Q9Z1A6 | |
Hdlbp | |
HDLBP Fusion Protein Ag7666 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |