Learn More
Kartell™ HDPE Narrow Neck Heavy-Walled Carboy Bottle
Store and dispense solutions and media using a HDPE carboy. Kartell™ HDPE Narrow Neck Heavy-Walled Carboy Bottle allows one to dispense most liquids, including reagents, distilled water and titrants.
2110.00 SEK - 2540.00 SEK
Specifications
Certifications/Compliance FDA
Description With O-ring seal cap
Autoclavable Yes
Includes Closure, Carrying Handles
Material HDPE
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10643852 View Documents Kartell™
0165000
50 L 2540.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
05-02-2020
Add to basket
10011061 View Documents Kartell™
0164800
25 L 2110.00 SEK
Pack of 4 View alternative products
Save Up to 
»
Estimated Shipment
07-02-2020
Add to basket
Description
Description
  • Very resistant
  • O-ring seal cap make it leakproof
  • The cap can be wired down for added safety
  • Includes carrying handles
  • Suitable for foodstuff
Specifications
Specifications
FDA
Yes
HDPE
Narrow
Heavy Walled Carboy
With O-ring seal cap
Closure, Carrying Handles
Suitable for food stuffs
Narrow