missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Heartland Virus Glycoprotein 2 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1619
Additional Details : Weight : 0.01000kg
Description
Heartland Virus Glycoprotein 2 Synthetic Peptide, PEP-1619, from Invitrogen.
This peptide corresponds to 19 amino acids near the center of Heartland Virus Glycoprotein 2.For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37ºC.Specifications
Virus | |
Heartland Virus Glycoprotein 2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
0.02% sodium azide | |
50 μg | |
PBS |
purified | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
No | |
Peptide | |
RUO - research use only |