Läs mer
Tocris Bioscience™ HEAT hydrochloride
GREENER_CHOICE
Produktkod. 18752494
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18752494 50 mg
18732784 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18752494 Leverantör Tocris Bioscience™ Leverantörsnummer 0535/50
Highly selective α1 antagonist
Very selective α1-adrenoceptor antagonist, precursor of the 3-[125I]-derivative.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30007-39-7
Molekylformel C19H22ClNO2
Specifikationer
Kemiskt namn eller material HEAT hydrochloride
Kvantitet 50 mg
Formel vikt 331.84
Renhet >98%
Molekylvikt (g/mol) 331.84