Learn More
Heating Elements For Cyclon Stills
Brand: Fistreem International WSC708.HEJ.4
3320.55 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
220V |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Heating Elements For Cyclon Stills > Voltage: 220V
Spot an opportunity for improvement?Share a Content Correction