Beskrivning
- Easily tackle heavy cleaning tasks - this multipurpose cleaning cloth is both strong and absorbent.
- Cut cleaning time by up to 35% and use up to 31% less effort than rental towels, as the cloth's suitable for repeated use.
- Protect hands from heat and metal scraps while you clean, thanks to this cleaning cloth's heavy-duty fabric.
- Cut down on your use of solvents by up to 41% and waste by up to 85% versus rags with the exelCLEAN™ material.
Specifikationer
Specifikationer
| Produkttyp | Heavy-duty Wiper |
| Längd (engelska) | 24.21 in. |
| Längd (metrisk) | 61.5 cm |
| Bredd (engelska) | 13.98 in. |
| Bredd (metrisk) | 35.5 cm |
| Färg | Vitt |
| Förpackningsmängd | 280/Cs. |
| Lager | 1-ply |