missing translation for 'onlineSavingsMsg'
Learn More

HECTD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20605-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840165

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
Specifications

Specifications

HECTD1
Polyclonal
Unconjugated
HECTD1
E3 ligase for inhibin receptor, EULIR, HECT domain containing 1, HECTD1, KIAA1131
Rabbit
Antigen Affinity Chromatography
RUO
207304, 25831
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q69ZR2, Q9ULT8
HECTD1
HECTD1 Fusion Protein Ag14581
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers