missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HECTD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20605-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.Specifications
HECTD1 | |
Polyclonal | |
Unconjugated | |
HECTD1 | |
E3 ligase for inhibin receptor, EULIR, HECT domain containing 1, HECTD1, KIAA1131 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
207304, 25831 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q69ZR2, Q9ULT8 | |
HECTD1 | |
HECTD1 Fusion Protein Ag14581 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |