Läs mer
Hederagenin, TRC
Produktkod. 30006036
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006036 10 mg
30007164 5 mg
30011868 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006036 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH12999010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 465-99-6
Molekylformel C30 H48 O4
Molekylvikt (g/mol) 472.7
Synonym (3β,4α)-3,23-Dihydroxy-olean-12-en-28-oic Acid, Caulosapogenin, Hederagenic Acid, Hederagenine, Hederagenol, NSC 24954;
IUPAC-namn (4aS,6aR,6aS,6bR,8aR,9R,10S,12aR,14bS)-10-hydroxy-9-(hydroxymethyl)-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
LEDER CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(=O)O
Specifikationer
Kemiskt namn eller material Caulosapogenin (Hederagenin)
Kvantitet 10 mg
InChI formel InChI=1S/C30H48O4/c1-25(2)13-15-30(24(33)34)16-14-28(5)19(20(30)17-25)7-8-22-26(3)11-10-23(32)27(4,18-31)21(26)9-12-29(22,28)6/h7,20-23,31-32H,8-18H2,1-6H3,(H,33,34)/t20-,21+,22+,23-,26-,27-,28+,29+,30-/m0/s1
Formel vikt 472.3553