Learn More
Heidolph™ Hei-TORQUE Expert Series Overhead Stirrer
Product Code. 15380648 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed:
10 to 2000 rpm
Model:
Hei-TORQUE Expert 100
Hei-TORQUE Expert 200
Hei-TORQUE Expert 400
Torque:
100 Ncm
200 Ncm
400 Ncm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Model Torque Unit Size
15380648 10 to 2000 rpm Hei-TORQUE Expert 200 200 Ncm Each
15391748 10 to 2000 rpm Hei-TORQUE Expert 100 100 Ncm Each
15321758 10 to 2000 rpm Hei-TORQUE Expert 400 400 Ncm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15380648 Supplier Heidolph™ Supplier No. 5016201000
  • ONLINE EXCLUSIVE PRICE
    26270.00 SEK
    25071.22 SEK / Paket med 1
    Save 1198.78 SEK
    4% Av
View all promotions
Beräknad leverans: 29-12-2026
Hei-TORQUE Expert Series Overhead Stirrers are designed to mix and disperse media that require non-reproducible results in high-viscosity applications.
  • Indication of torque tendencies to monitor viscosity changes
  • Digital 2.4 in. display for ease of operation
  • Constant speed even under changing loads
  • Safe start and stop of operation via slide touch panel to avoid unintended stirring
  • Newest motor generation for maximum power at minimum noise level
TRUSTED_SUSTAINABILITY
Specifications
Speed 10 to 2000 rpm
Capacity 50 L
Display Digital
Voltage 230 V
Wattage 120 W
Diameter (English) Shaft 0.4 in.
Diameter (Metric) Shaft 10.5 mm
Dimensions (L x W x H) 241 x 86 x 328 mm
Model Hei-TORQUE Expert 200
No. of Speed Gears 1
Protection Class IP54
Torque 200 Ncm
Type Overhead Stirrer
Viscosity 100,000 mPas
Weight (English) 11.9 lb.
Weight (Metric) 5.4 kg