missing translation for 'onlineSavingsMsg'
Learn More

HEK 293T cells Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27555-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825695

  • 1656.73 SEK / 20µL
Estimated Shipment: 23-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is designed to detect the proteins of the HEK 293T cells.

HEK 293T cell line, an important variant of HEK 293 cell line, which contains the SV40 Large T-antigen that allows for episomal replication of transfected plasmids containing the SV40 origin of replication. HEK 293T cells are usually used for plasmids transfection and temporal expression of desired gene products.
Specifications

Specifications

HEK 293T cells
Polyclonal
Unconjugated
293T, HEK 293T cells, HEK293T
Protein A
RUO
Human
Antibody
IgG
ELISA
1.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
20 μL
Primary
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers