Läs mer
Heliomethylamine Hydrochloride, TRC
Produktkod. 30407964
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30407964 500 mg
1 options
Produktkod. 30407964 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH235100500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Heliomethylamine Hydrochloride
CAS 1797883-86-3
Molekylformel C12 H17 N O2 . Cl H
Kvantitet 500 mg
Synonym 1,3-Benzodioxole-5-propanamine, N,β-dimethyl-, hydrochloride (1:1), Heliomethylamine hydrochloride, 3,4-MDMA methylene homolog hydrochloride, 3-(1,3-Benzodioxol-5-yl)-N,2-dimethylpropan-1-amine hydrochloride
InChI formel InChI=1S/C12H17NO2.ClH/c1-9(7-13-2)5-10-3-4-11-12(6-10)15-8-14-11;/h3-4,6,9,13H,5,7-8H2,1-2H3;1H
LEDER Cl.CNCC(C)Cc1ccc2OCOc2c1
IUPAC-namn 3-(1,3-benzodioxol-5-yl)-N,2-dimethylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 243.73
Formel vikt 243.1026